missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Borosilicate Glass Test Tube
Store test reagents and materials. Fisherbrand™ borosilicate glass test tubes come in a variety of sizes.
Brand: Fisherbrand™
45.40 GBP valid until 2024-04-30
Use promo code "21749" to get your promotional price.
Additional Details : Weight : 0.54000kg
Description
- Borosilicate glass test tubes, grade 3.3
- All ISO 4142
Specifications
ISO 4142 | |
Borosilicate Glass | |
12 mm |
75 mm | |
100 Pack |