missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Borosilicate Glass Squat Form Beakers
Click to view available options
Capacity (Metric):
5 mL
10 mL
Description
- With spout
- Graduated
- Borosilicate glass
Specifications
Specifications
| Capacity (Metric) | 10 mL |
| Material | Borosilicate Glass |
| Includes | Spout |
| Quantity | 10 Pk. |
| Type | Beaker |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction