missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences DURAN™ Protect Clear Borosilicate Glass Laboratory Bottles with Plastic Safety Coating, DIN Thread, Graduations, Bottle Only

Product Code. 11669908 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
100 mL
150 mL
250 mL
500 mL
750 mL
1000 mL
2000 mL
3500 mL
5000 mL
10 L
15 L
20 L
Unit Size:
Each
Pack of 10
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
11669908 3500 mL Each
11762312 100 mL Pack of 10
11782312 500 mL Pack of 10
11792312 1000 mL Pack of 10
10051192 25 mL Pack of 10
10041192 50 mL Pack of 10
11772312 250 mL Pack of 10
10098060 5000 mL Each
11702322 2000 mL Pack of 10
10625131 10 L Each
11659828 20 L Each
11689898 150 mL Pack of 10
11629908 750 mL Pack of 10
13197216 15 L Each
13417119 10 mL Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
This item is not returnable. View return policy
Product Code. 11669908 Supplier DWK Life Sciences Supplier No. 218056903

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

DURAN™ Protect Laboratory Bottles are clear, multifunctional borosilicate glass laboratory bottles with a protective safety coating, DIN Threads, and graduations.

  • DURAN™ Original GL laboratory bottles have become the gold standard of multifunctional laboratory bottles since their introduction in 1972.
  • The protect safety-coated version of the bottles combining the best of both worlds: virtually inert USP/EP Type I borosilicate glass 3.3 glass on the inside and a protective safety plastic layer on the outside.
  • The bottles are moulded from high-purity, clear, glass that is then coated with a clear PU plastic coating to reduce the hazards of broken glass and liquid splashes in the event of breakage.
  • A combination of high-performance materials and robust ISO 4796-1 design provides a long working life.
  • The bottles feature an easy-to-read graduated volume scale, and marking field in permanent white enamel.
  • Permanent Retrace Code on each bottle for manufacturing lot traceability.
  • The protect safety coating meets the requirements of ASTM E1522, and is fully autoclavable at 121 or 134°C.
  • The coating provides scratch, leak* and splinter* protection (* only applies to bottles 5000 mL and less).
  • Highly resistance, long service-life coating.
  • Standardised GL-thread and matching cap system for particularly tight sealing and simple, clean pouring out.
  • UV protection up to approx. 380 nm due to external coating.
  • Easy-to-read scale with graduation marks to estimate the content.
  • Fired-on, highly durable white ceramic print.
  • Sizes 10 and 25 mL with specially shaped glass edge for improved pouring out whereby an additional plastic pouring ring is not required.
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 3500 mL
Capacity (English) 0.9 gal.
Coating Polyurethane
Color Clear
Diameter (English) 6.3 in.
For Use With (Application) Autoclaving of media and reagents; Preparation of media and reagents; Sample collection; Culturing vessel; Storage of media and reagents; Transport and shipping; Liquid handling reservoir
Material Borosilicate Glass (3.3)
Quantity 1 Ea.
Thread Size (Metric) GL 45
Certifications/Compliance ASTM E438, Type I, Class A
Closure Included No
Disposable Reusable
Graduated Yes
Mouth Narrow
Shape Round
Height (Metric) 295 mm
Product Type Laboratory Bottle
Height (English) 11.6 in.
Closure Size 45 mm
Diameter (Metric) 160 mm
Autoclavable Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.