missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Manufactured from borosilicate glass with a heavy wall design for strong resistance to heat and chemicals. Fisherbrand™ Borosilicate Glass Heavy Wall Rimmed Test Tubes are ideal for chemical and biological work.
- Features: rimmed heavy walls, superior mechanical strength
- Grade: 3.3 borosilicate glass test tubes
- Complies to: ISO 4142
Specifications
Specifications
| Certifications/Compliance | ISO 4142 |
| Length (Metric) | 125 mm |
| Material | Borosilicate Glass |
| Quantity | 100 Pack |
| Rimmed | Yes |
| Diameter (Metric) | 16 mm |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?