missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Robu™ Borosilicate Glass Buchner Filter Funnels
Brand: Robu™ 21504
Additional Details : Weight : 0.10000kg
Specifications
40mm | |
40mm (OD); 36mm (ID) | |
4 | |
50 mL | |
Borosilicate Glass |
10mm | |
Highly resistant to most chemicals such as solvents, acids and alkalis | |
Buchner filter funnel | |
53 mm |