Learn More
DWK Life Sciences™ DURAN™ Vacuum Desiccator, with NOVUS standard ground joint (24/29) junction tube in the lid, with stopcock, with flat flange
Product Code. 10291361 Shop All DWK Life Sciences Products
Change view
Click to view available options
Volume (Metric):
700 mL
2.4 L
5.8 L
10.5 L
18.5 L
Description:
With porcelain plate
Without porcelain plate
Unit Size:
Each
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Description unitSize
10291361 5.8 L Without porcelain plate Each
10221321 700 mL Without porcelain plate Each
10231321 2.4 L Without porcelain plate Each
10129800 10.5 L Without porcelain plate Each
10549051 18.5 L Without porcelain plate Each
13465509 18.5 L With porcelain plate Each
13495499 10.5 L With porcelain plate Each
13435499 5.8 L With porcelain plate Each
13405499 2.4 L With porcelain plate Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 10291361 Supplier DWK Life Sciences™ Supplier No. 247826108
Only null left
Add to basket
Add to basket
The desiccator is used to store moisture-sensitive substances and to dry moist products. The NOVUS design has a standard ground joint (NS 24/29) junction tube in the lid.
The DURAN™ desiccator is a vacuum-suitable laboratory vessel with a ground lid, which is used for storing moisture-sensitive substances and for drying moist products. Separated by a sieve plate (made of metal or porcelain), a drying agent is introduced into the lower part of the desiccator base, which binds the moisture of the substance to be dried, placed on the sieve plate. To accelerate the drying process, a vacuum can be connected if necessary, as the liquid evaporates faster due to a lower pressure in the desiccator. The vacuum is applied via a shut-off valve connection. To facilitate loosening of the lid, the surface grinding of the desiccator must be well greased, since after evacuation the lid is pressed onto the lower part. All DURAN™ desiccators are vacuum-resistant, i.e. they can be used up to the technically maximum possible vacuum.
  • Due to the high wall-thickness of the vessels and the exact machining of the vacuum-tight ground joints on the lid and base, storage under vacuum is possible – even over extremely long periods
  • To accelerate the drying process, a vacuum can be applied via the stopcock
  • Suitable for full vacuum (0 mbar)
  • Spare parts such as lids, bases, stopcocks and caps can be interchanged as required (observe DN)
  • Available with or without porcelain plate
TRUSTED_SUSTAINABILITY
Specifications
Description Without porcelain plate
Material Borosilicate Glass 3.3, PTFE
Product Type Vacuum Dessicator
For Use With (Application) Ideal for removing moisture from solids; Storage of moisture-sensitive materials over long periods; For applications requiring vacuum desiccation; Drying of materials; Safe working under vacuum
Quantity 1 Each
Volume (Metric) 5.8 L