Learn More
DWK Life Sciences™ DURAN™ Desiccator Lid, with threaded outlet, type MOBILEX (GL 32), suitable for all types of bases
Product Code. 10085220 Shop All DWK Life Sciences Products
Change view
Click to view available options
Description:
DN 150, GL 32
DN 200, GL 32
DN 250, GL 32
DN 300, GL 32
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description unitSize
10085220 DN 200, GL 32 Each
10481591 DN 250, GL 32 Each
10340311 DN 150, GL 32 Each
10095220 DN 300, GL 32 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10085220 Supplier DWK Life Sciences™ Supplier No. 244406105
Only null left
Add to basket
Add to basket
The desiccator lid is used to cover or seal the desiccator base.
The DURAN™ desiccator is a vacuum-suitable laboratory vessel with a ground lid, which is used for storing moisture-sensitive substances and for drying moist products. Separated by a sieve plate (made of metal or porcelain), a drying agent is introduced into the lower part of the desiccator base, which binds the moisture of the substance to be dried, placed on the sieve plate. To accelerate the drying process, a vacuum can be connected if necessary, as the liquid evaporates faster due to a lower pressure in the desiccator. The vacuum is applied via a shut-off valve connection. To facilitate loosening of the lid, the surface grinding of the desiccator must be well greased, since after evacuation the lid is pressed onto the lower part. All DURAN™ desiccators are vacuum-resistant, i.e. they can be used up to the technically maximum possible vacuum.
  • Suitable for all types of bases
  • MOBILEX type with GL 32 thread on the lid
TRUSTED_SUSTAINABILITY
Specifications
Description DN 200, GL 32
Material Borosilicate Glass (3.3)
For Use With (Application) Ideal for removing moisture from solids; Storage of moisture-sensitive materials over long periods; For applications requiring vacuum desiccation; Drying of materials; Safe working under vacuum