missing translation for 'onlineSavingsMsg'
Learn More

BOP1 Antibody (T7-P1B7*A8) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

£159.00 - £302.00

Specifications

Antigen BOP1
Clone T7-P1B7*A8
Concentration 1 mg/mL
Dilution Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1/3, Immunohistochemistry-Paraffin 1/3
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18674138
View Documents
Novus Biologicals
NBP2-50124-0.025ml
0.025 mL
£159.00
0.03mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18606126
View Documents
Novus Biologicals
NBP2-50124
0.1 mL
£302.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

BOP1 Monoclonal antibody specifically detects BOP1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications

Specifications

BOP1
1 mg/mL
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Epigenetics
PBS
23246
IgG1 κ
Protein A purified
T7-P1B7*A8
Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1/3, Immunohistochemistry-Paraffin 1/3
Monoclonal
Purified
RUO
Human
block of proliferation 1, KIAA0124Block of proliferation 1 protein, ribosome biogenesis protein BOP1
Peptide Sequence - KGHVLTRDLG
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.