missing translation for 'onlineSavingsMsg'
Learn More

BOP1 Antibody (T7-P1B7*A8) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-50124-0.025ml

 View more versions of this product

Product Code. 18674138

  • £159.00 / 0.03mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

BOP1 Monoclonal antibody specifically detects BOP1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

BOP1
Monoclonal
1 mg/mL
Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1/3, Immunohistochemistry-Paraffin 1/3
block of proliferation 1, KIAA0124Block of proliferation 1 protein, ribosome biogenesis protein BOP1
Peptide Sequence - KGHVLTRDLG
0.025 mL
Epigenetics
23246
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1 κ
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
T7-P1B7*A8
Unconjugated
PBS
Mouse
Protein A purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.