missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BODIPY™ FL Maleimide (BODIPY™ FL N-(2-Aminoethyl))Maleimide)

Product Code. 11584137
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11584137 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 11584137 Supplier Invitrogen™ Supplier No. B10250
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

BODIPY™ FL Maleimide (BODIPY™ FL N-(2-Aminoethyl))Maleimide)

The thiol-reactive BODIPY™ FL maleimide produces electronically neutral dye conjugates that are spectrally similar to the negatively charged fluorescein dye. This dye's lack of ionic charge results in minimal effects on the isoelectric points of standard proteins conjugated with this fluorophore. The small size and relatively long excited-state lifetime of BODIPY™ FL dye has proven useful for studying ligand-receptor interaction by fluorescence polarization. In addition, BODIPY™ FL dye has little or no spectral overlap with longer-wavelength dyes such as tetramethylrhodamine and Texas Red™ dye, making it a useful green fluorophore for multicolor applications.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 mg
Product Type Maleimide
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Chemical Reactivity Thiol
Shipping Condition Room Temperature
Reactive Moiety Maleimide
Label or Dye BODIPY™ FL
Label Type BODIPY Dyes
Product Line BODIPY

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.