missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MedChemExpress Boc-NH-PEG3
Click to view available options
:
100 mg
Specifications
Specifications
| Molecular Weight (g/mol) | 249.3 g/mol |
| CAS | 139115-92-7 |
| Content And Storage | Pure form -20°C 3 years, 4°C 2 years ∣ In solvent -80°C 6 months, -20°C 1 month |
| Shipping Condition | Room temperature |
| Quantity | 100 mg |
| Synonym | PROTAC Linker 10 |
| Target | NO Synthase ∣ MMP ∣ Endogenous Metabolite |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction