missing translation for 'onlineSavingsMsg'
Learn More

BOC-N-methyl-D-alanine, 97%, Thermo Scientific™

Product Code. 10341750 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10341750 5 g 5g
10491240 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10341750 Supplier Thermo Scientific Acros Supplier No. 434240050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 19914-38-6
Molecular Formula C9H17NO4
Molecular Weight (g/mol) 203.24
MDL Number MFCD00063137
InChI Key VLHQXRIIQSTJCQ-ZCFIWIBFSA-N
Synonym boc-n-methyl-d-alanine, boc-n-me-d-ala-oh, n-boc-n-methyl-d-alanine, boc-d-n-me-ala-oh, boc-n-alpha-methyl-d-alanine, r-2-tert-butoxycarbonyl methyl amino propanoic acid, n-tert-butoxycarbonyl-n-methyl-d-alanine, d-alanine, n-1,1-dimethylethoxy carbonyl-n-methyl, 2r-2-tert-butoxycarbonyl methyl amino propanoic acid
PubChem CID 16211306
IUPAC Name (2R)-2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]propanoic acid
SMILES CC(C(=O)O)N(C)C(=O)OC(C)(C)C

Spécification

Melting Point 92.3°C to 94°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
Specific Rotation 29° to 31° (20°C, 589nm) (c=1, ethanol)
Formula Weight 203.24
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material BOC-N-Methyl-D-alanine

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.