CAS RN 19914-38-6
IUPAC Name:
2-{[(tert-butoxy)carbonyl](methyl)amino}propanoic acid
Synonyms:
2-[(tert-butoxycarbonyl)(methyl)amino]propanoic acid
Molecular Weight (g/mol):
203.24
Molecular Formula:
C9H17NO4
InChi Key:
VLHQXRIIQSTJCQ-UHFFFAOYNA-N
SMILES:
CC(N(C)C(=O)OC(C)(C)C)C(O)=O
1
–
1
of
1
results
1
BOC-N-methyl-D-alanine, 97%, Thermo Scientific™
CAS: 19914-38-6 Molecular Formula: C9H17NO4 Molecular Weight (g/mol): 203.24 MDL Number: MFCD00063137 InChI Key: VLHQXRIIQSTJCQ-ZCFIWIBFSA-N Synonym: boc-n-methyl-d-alanine,boc-n-me-d-ala-oh,n-boc-n-methyl-d-alanine,boc-d-n-me-ala-oh,boc-n-alpha-methyl-d-alanine,r-2-tert-butoxycarbonyl methyl amino propanoic acid,n-tert-butoxycarbonyl-n-methyl-d-alanine,d-alanine, n-1,1-dimethylethoxy carbonyl-n-methyl,2r-2-tert-butoxycarbonyl methyl amino propanoic acid PubChem CID: 16211306 IUPAC Name: (2R)-2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]propanoic acid SMILES: CC(C(=O)O)N(C)C(=O)OC(C)(C)C