missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Boc-C1-PEG2-C4-Cl

Product Code. 16405156
Encompass_Preferred
Change view
Click to view available options
:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16405156 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16405156 Supplier MedChemExpress Supplier No. HY108371

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Boc-C1-PEG2-C4-Cl (PROTAC Linker 1) is a PEG-based PROTAC linker can be used in the synthesis of PROTACs.

Specifications

Molecular Weight (g/mol) 294.81 g/mol
CAS 1835705-53-7
Content And Storage Pure form -20°C 3 years, 4°C 2 years ∣ In solvent -80°C 6 months, -20°C 1 month
Shipping Condition Room temperature
Quantity 100 mg
Synonym PROTAC Linker 1
Solubility DMSO : ≥ 50 mg/mL (169.60 mM)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.