missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMSC UbP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | BMSC UbP |
|---|---|
| Concentration | 0.17 mg/mL |
| Applications | Immunocytochemistry, Immunofluorescence, Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16838733
|
Proteintech
11564-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16828733
|
Proteintech
11564-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
UBL7 (Ubiquitin Like 7) is a Protein Coding gene.Specifications
| BMSC UbP | |
| Immunocytochemistry, Immunofluorescence, Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Rat, Human, Mouse | |
| Q91W67, Q96S82 | |
| 300744, 69459, 84993 | |
| BMSC UbP Fusion Protein Ag2130 | |
| Primary | |
| -20°C | |
| UBL7 |
| 0.17 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| BMSC UbP, BMSCUBP, TCBA1, Ubiquitin like protein 7, Ubiquitin like protein SB132, UBL7 | |
| UBL7 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title