Learn More
BMSC UbP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11564-1-AP-150UL
 View more versions of this product
Product Code. 16828733
  • £349.00 / 150µL
Description
UBL7 (Ubiquitin Like 7) is a Protein Coding gene.
Specifications
Specifications
BMSC UbP
Polyclonal
Unconjugated
UBL7
BMSC UbP, BMSCUBP, TCBA1, Ubiquitin like protein 7, Ubiquitin like protein SB132, UBL7
Rabbit
Antigen Affinity Chromatography
RUO
300744, 69459, 84993
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.17 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q91W67, Q96S82
UBL7
BMSC UbP Fusion Protein Ag2130
150 μL
Primary
Human, Mouse, Rat