missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMP-8b Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£200.00 - £558.00
Specifications
| Antigen | BMP-8b |
|---|---|
| Dilution | Western Blot 1:500-1:1000, ELISA 1:2000 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18279454
|
Novus Biologicals
NB300-698-0.025MG |
0.025 mg |
£200.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18252342
|
Novus Biologicals
NB300-698-0.05MG |
0.05 mg |
£341.00
0.05mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18119039
|
Novus Biologicals
NB300-698 |
0.1 mg |
£558.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BMP-8b Polyclonal specifically detects BMP-8b in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.Specifications
| BMP-8b | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rat, Human, Mouse | |
| 656 | |
| Synthetic peptide: LQTLRAGDEGWLVL conjugated to KLH, corresponding to amino acids 184-197 of BMP8. | |
| Primary |
| Western Blot 1:500-1:1000, ELISA 1:2000 | |
| Polyclonal | |
| Rabbit | |
| BMP-8, bone morphogenetic protein 8b, MGC131757 | |
| BMP8B | |
| IgG | |
| BMP8 |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title