missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMP-8b Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB300-698
This item is not returnable.
View return policy
Description
BMP-8b Polyclonal specifically detects BMP-8b in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.
Specifications
| BMP-8b | |
| Polyclonal | |
| Unconjugated | |
| Tris/Glycine buffer, pH 7.4-7.8, HEPES,BSA 0.5%, glycerol 30%. with 0.02% Sodium Azide | |
| BMP8B | |
| Synthetic peptide: LQTLRAGDEGWLVL conjugated to KLH, corresponding to amino acids 184-197 of BMP8. | |
| 0.1 mg | |
| Cytokine Research | |
| 656 | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 1:500-1:1000, ELISA 1:2000 | |
| BMP-8, bone morphogenetic protein 8b, MGC131757 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| BMP8 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction