missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMP-4 Antibody (3C11H8) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52423-0.025mg
This item is not returnable.
View return policy
Description
BMP-4 Monoclonal specifically detects BMP-4 in Human, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, CyTOF-ready.
Specifications
| BMP-4 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:100-1:500, CyTOF-ready | |
| BMP-2B, BMP2B1, BMP2BMCOPS6, BMP-4, Bone morphogenetic protein 2B, bone morphogenetic protein 4, DVR4, OFC11, ZYME | |
| Mouse | |
| 46.5 kDa | |
| 0.025 mg | |
| Biologically Active Proteins, Embryonic Stem Cell Markers, Stem Cell Markers, Stem Cell Signaling Pathway | |
| 652 | |
| Human, Rat | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, CyTOF | |
| 3C11H8 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| BMP4 | |
| Purified recombinant fragment of human BMP4 (AA: 277-408) expressed in E. Coli. | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction