missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMP-4 Antibody (3C11H8) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£161.00 - £435.00
Specifications
| Antigen | BMP-4 |
|---|---|
| Clone | 3C11H8 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:100-1:500, CyTOF-ready |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18231030
|
Novus Biologicals
NBP2-52423 |
0.1 mg |
£435.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18665448
|
Novus Biologicals
NBP2-52423-0.025mg |
0.025 mg |
£161.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BMP-4 Monoclonal specifically detects BMP-4 in Human, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, CyTOF-ready.Specifications
| BMP-4 | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, CyTOF | |
| Unconjugated | |
| Mouse | |
| Biologically Active Proteins, Embryonic Stem Cell Markers, Stem Cell Markers, Stem Cell Signaling Pathway | |
| PBS with 0.05% Sodium Azide | |
| 652 | |
| Purified recombinant fragment of human BMP4 (AA: 277-408) expressed in E. Coli. | |
| Primary | |
| 46.5 kDa |
| 3C11H8 | |
| Western Blot 1:500-1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:100-1:500, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Rat | |
| BMP-2B, BMP2B1, BMP2BMCOPS6, BMP-4, Bone morphogenetic protein 2B, bone morphogenetic protein 4, DVR4, OFC11, ZYME | |
| BMP4 | |
| IgG1 | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title