missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMP-2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£240.00 - £409.00
Specifications
| Antigen | BMP-2 |
|---|---|
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18283312
|
Novus Biologicals
NBP2-56251 |
100 μL |
£409.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18621459
|
Novus Biologicals
NBP2-56251-25ul |
25 μL |
£240.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BMP-2 Polyclonal specifically detects BMP-2 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| BMP-2 | |
| Polyclonal | |
| Rabbit | |
| Cytokine Research, Embryonic Stem Cell Markers, Immunology, Neuronal Cell Markers, Neuronal Stem Cell Markers, Stem Cell Markers, Stem Cell Signaling Pathway | |
| BMP-2, BMP-2A, BMP2ABone morphogenetic protein 2A, bone morphogenetic protein 2 | |
| BMP2 | |
| IgG | |
| Affinity Purified |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 650 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:RLVNQNASRWESFDVTPAVMRWTAQGHANHGFVVEVAHLEEKQGVSKRHVRIS | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title