missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMP-15/GDF-9B Rabbit anti-Human, Mouse, Rat, Clone: 6K5C3, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | BMP-15/GDF-9B |
|---|---|
| Clone | 6K5C3 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18395585
|
Novus Biologicals
NBP3-16342-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18308554
|
Novus Biologicals
NBP3-16342-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BMP-15/GDF-9B Monoclonal antibody specifically detects BMP-15/GDF-9B in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| BMP-15/GDF-9B | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis, Cardiovascular Biology, Cell Biology, Cytokines and Growth Factors, Immunology, Signal Transduction, Stem Cells | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 9210 | |
| IgG | |
| Affinity purified |
| 6K5C3 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| BMP-15, bone morphogenetic protein 15, GDF-9B, Growth/differentiation factor 9B, ODG2, POF4 | |
| A synthetic peptide corresponding to a sequence within amino acids 293-392 of human BMP-15/GDF-9B (O95972). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title