missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMP-15/GDF-9B Rabbit anti-Human, Mouse, Rat, Clone: 6K5C3, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-16342-100UL
This item is not returnable.
View return policy
Description
BMP-15/GDF-9B Monoclonal antibody specifically detects BMP-15/GDF-9B in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| BMP-15/GDF-9B | |
| Monoclonal | |
| Unconjugated | |
| BMP-15, bone morphogenetic protein 15, GDF-9B, Growth/differentiation factor 9B, ODG2, POF4 | |
| A synthetic peptide corresponding to a sequence within amino acids 293-392 of human BMP-15/GDF-9B (O95972). | |
| 100 μg | |
| Angiogenesis, Cardiovascular Biology, Cell Biology, Cytokines and Growth Factors, Immunology, Signal Transduction, Stem Cells | |
| 9210 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| 6K5C3 | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction