missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMAL1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£168.00 - £356.00
Specifications
| Antigen | BMAL1 |
|---|---|
| Dilution | Western Blot 1:100 - 1:200, Simple Western 1:100, Chromatin Immunoprecipitation (ChIP) 1:10-1:500 |
| Applications | Western Blot, ChIP Assay |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18083704
|
Novus Biologicals
NB100-129SS |
0.1 mL |
£168.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18745243
|
Novus Biologicals
NB100-129 |
0.4 mL |
£356.00
0.40mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BMAL1 Polyclonal specifically detects BMAL1 in Human samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation.Specifications
| BMAL1 | |
| Western Blot, ChIP Assay | |
| Unconjugated | |
| Rabbit | |
| Cholesterol Metabolism, Chromatin Research, Circadian Rhythm, Lipid and Metabolism, Neuroscience, Transcription Factors and Regulators | |
| Whole antisera with 0.02% Sodium Azide | |
| aryl hydrocarbon receptor nuclear translocator-like, aryl hydrocarbon receptor nuclear translocator-like protein 1, basic-helix-loop-helix-PAS orphan MOP3, BHLHE5, bHLHe5brain and muscle, bHLH-PAS protein JAP3, BMAL1TIC, Brain and muscle ARNT-like 1, Class E basic helix-loop-helix protein 5, Member of PAS protein 3, member of PAS superfamily 3, MOP3BMAL1c, PAS domain-containing protein 3, PASD3MGC47515 | |
| ARNTL | |
| IgG | |
| Unpurified | |
| 73 kDa |
| Western Blot 1:100 - 1:200, Simple Western 1:100, Chromatin Immunoprecipitation (ChIP) 1:10-1:500 | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Human | |
| O00327 | |
| 406 | |
| Recombinant protein corresponding to residues 294-624 of human BMAL1. [UniProt# O00327] | |
| Primary | |
| Store at 4C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title