missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMAL1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-129
This item is not returnable.
View return policy
Description
BMAL1 Polyclonal specifically detects BMAL1 in Human samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation.
Specifications
| BMAL1 | |
| Polyclonal | |
| Western Blot 1:100 - 1:200, Simple Western 1:100, Chromatin Immunoprecipitation (ChIP) 1:10-1:500 | |
| O00327 | |
| ARNTL | |
| Recombinant protein corresponding to residues 294-624 of human BMAL1. [UniProt# O00327] | |
| Unpurified | |
| RUO | |
| Primary | |
| Human | |
| Antisera |
| Western Blot, ChIP Assay | |
| Unconjugated | |
| Whole antisera with 0.02% Sodium Azide | |
| aryl hydrocarbon receptor nuclear translocator-like, aryl hydrocarbon receptor nuclear translocator-like protein 1, basic-helix-loop-helix-PAS orphan MOP3, BHLHE5, bHLHe5brain and muscle, bHLH-PAS protein JAP3, BMAL1TIC, Brain and muscle ARNT-like 1, Class E basic helix-loop-helix protein 5, Member of PAS protein 3, member of PAS superfamily 3, MOP3BMAL1c, PAS domain-containing protein 3, PASD3MGC47515 | |
| Rabbit | |
| 73 kDa | |
| 0.4 mL | |
| Cholesterol Metabolism, Chromatin Research, Circadian Rhythm, Lipid and Metabolism, Neuroscience, Transcription Factors and Regulators | |
| 406 | |
| Store at 4C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction