Learn More
Agilent BioTek Blue Test Dye, 10X Concentrate
GREENER_CHOICE
Product Code. 10586360 Shop All Agilent BioTek Products
Click to view available options
Quantity:
125 mL
Unit Size:
125mL
Product Code. 10586360
Brand: Agilent BioTek 7773001
For use with ELx405 and ELx50 Microplate Washers
  • Used for Qualification Tests
  • For use with ELx405 and ELx50 Microplate Washers
TRUSTED_SUSTAINABILITY
Specifications
Product Type Blue Test Dye, 10X Concentrate
For Use With (Equipment) Agilent BioTek Microplate Washers and Readers
Quantity 125 mL