Learn More
3M™ Blue SMS Based Fabric Category III Protective Coveralls
Product Code. p-8003511
Click to view available options
Size:
4X-Large
Unit Size:
Pack of 20
Product Code. 11983764
Brand: 3M™ 7000089624
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Product Type General Purpose Coveralls
Size 4X-Large
Gender Unisex
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type 2-panel
Seam Type Serged
Sterility Non-sterile
Protection Class Category III
Height (Metric) ‎200 to 206 cm