Learn More
3M™ Protective Coverall 4532+, Blue
Product Code. 15750048
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Unit Size:
Pack of 20
Product Code. 15750048
Brand: 3M™ 7100016004
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Coverall has a unique specialist material treatment which offers enhanced liquid hold out with added oil and alcohol repellency. Certified to Type 5 for hazardous particles and Type 6 for limited chemical splash, 4532+ coverall offers the soft breathable comfort of an SSMMS with the improved liquid performance normally associated with a laminate.
  • Unique liquid repellency with oil and alcohol resistance material treatment
  • Soft and strong multi-layered 52 gsm SSMMS for strong performance
  • Made from super breathable SSMMS non-woven for effective release of body heat and moisture and reduced heat stress
  • 3 panel hood, reinforced triangular gusset panel, 2-way zip and storm flap with sealable tape, elasticated back and ankles to better freedom of movement
  • Anti-static coating EN 1149-5 reduces static discharge and the risk of sparks
  • EN 1073-2 nuclear particulate hold out
  • Knitted cuffs and elastic waist and ankles for a better fit and freedom of movement
TRUSTED_SUSTAINABILITY
Specifications
Product Type Protective Coveralls
Size 4X-Large
Gender Unisex
Material SSMMS
Color Blue
Disposable Single-use
Waist Style Elastic
Cuff Style Knit
Closure Type Front Zipper
Sterility Non-sterile
Certifications/Compliance CE Category III , EN 1073-2 radioactive particles , EN 1149-5 - electrostatic properties , EN 13034: 2005 + A1: 2009 Type 6 liquid splash , EN ISO 13982-1: 2004 + A1: 2010 Type 5 harmful particles
Certifications/Compliance (Europe) CE Category III, EN 1073-2, EN 1149-5, EN 13034:2005+A1:2009 Type 6, EN ISO 13982-1:2004 Type 5