missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Blue Measuring Spoon
GREENER_CHOICEBIOPHARMA

Product Code. 16595240 Shop All Bürkle Products
Click to view available options
Capacity (Metric):
0.5 mL
1 mL
2.5 mL
5 mL
10 mL
15 mL
25 mL
50 mL
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 16595240

Brand: Buerkle™ 53784055

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Measuring spoons in a unique shade of blue that does not occur often in foodstuffs, making them easy to find and visually identify.

  • Sterile versions are individually packaged and radiation sterilized
  • The use of blue tools thus helps to meet the requirements of the HACCP hazard analysis and assessment in reference to foreign objects
  • This is an important criterion, especially in food processing and production and in the animal feed and pharmaceutical industry
  • Volumetric and dosing spoons can be used to measure small amounts of powder, granules and liquids exactly and dose them precisely
  • For precise measuring, excess medium is stripped off the upper edge of the spoon
  • The content volume is engraved on the handle
  • The flat front edge of the volumetric spoon is ideal for picking up medium from the bottom of the container or for getting into the corners of the container, thus allowing for good emptying of small remaining amounts
  • The flat base makes it possible to set the volumetric spoon down without spilling the content
  • The disposable dosing spoon is suitable for use as a sample spoon for taking small sample quantities
  • Since the plastic volumetric spoon was developed for single use, there is no cleaning and the possibility of cross-contamination and impurities is excluded
  • The dosing spoons and volumetric spoons are suitable for precise measuring for chemical and pharmaceutical applications, cosmetics, animal feed and the food and beverage industry
  • Can be used as part of HACCP/IFS foreign object management
  • Especially for the food, animal feed and pharmaceutical industries
  • With stripper edge
  • For single use
  • Cleanroom manufactured (Class 7)
  • Complies with EU food requirements and FDA regulations
TRUSTED_SUSTAINABILITY

Specifications

Quantity 100
Description Dosing spoons with stripper edge for exact measuring
Sterility Sterile
Capacity (Metric) 15 mL
Color Blue
Certifications/Compliance Food Contact, Reach, RoHS
Disposable Single-use
Product Type Spoon
For Use With (Application) Especially for The Food, Animal Feed and Pharmaceutical Industries
Material Polystyrene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.