missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Blue Caps for Rimless Test Tubes

Product Code. 11562652 Shop All Fisherbrand Products
Change view
Click to view available options
Diameter (Metric) Outer:
19 mm
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outer unitSize
11562652 19 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11562652
Only null left
Add to basket
Add to basket

Closures for rimless test tubes.

  • Bacti-Caps™ provide a firm, positive culture tube closure allowing a controlled gas interchange, thus proving a more practical alternative to cotton wool plugs
  • Fit rimless and plastic tubes with an outside diameter of 19mm
  • The three flexible fins moulded on the inner wall of each cap ensure a positive fit even when minor variations in tube size occur
  • Made from chemically resistant virgin plastic so they are unaffected by culture media, most acids, alkalis, alcohols and esters

Specifications

Diameter (Metric) Outer 19 mm
Product Type Culture Tube Closure
Color Blue
Quantity 100 Pack
For Use With (Equipment) Rimless Test Tubes with an outside diameter of 19mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.