missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Blood group H inhibitor Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£388.00
Specifications
| Antigen | Blood group H inhibitor |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
Blood group H inhibitor Polyclonal specifically detects Blood group H inhibitor in Human samples. It is validated for Western Blot.Specifications
| Blood group H inhibitor | |
| Polyclonal | |
| Rabbit | |
| P19526 | |
| 2523 | |
| IgG | |
| 41 kDa |
| Western Blot | |
| Unconjugated | |
| RUO | |
| alpha (1,2) fucosyltransferase, Alpha(1,2)FT 1,2-alpha-L-fucosyltransferase, Blood group H alpha 2-fucosyltransferase, EC 2.4.1.69, Fucosyltransferase 1, fucosyltransferase 1 (galactoside 2-alpha-L-fucosyltransferase), fucosyltransferase 1 (galactoside 2-alpha-L-fucosyltransferase, Bombayphenotype included), fucosyltransferase 1 (galactoside 2-alpha-L-fucosyltransferase, H blood group), galactoside 2-alpha-L-fucosyltransferase 1, GDP-L-fucose:beta-D-galactoside 2-alpha-L-fucosyltransferase 1, HHH, HSC | |
| FUT1 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title