missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Blood group H inhibitor Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-57016
This item is not returnable.
View return policy
Description
Blood group H inhibitor Polyclonal specifically detects Blood group H inhibitor in Human samples. It is validated for Western Blot.
Specifications
| Blood group H inhibitor | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| alpha (1,2) fucosyltransferase, Alpha(1,2)FT 1,2-alpha-L-fucosyltransferase, Blood group H alpha 2-fucosyltransferase, EC 2.4.1.69, Fucosyltransferase 1, fucosyltransferase 1 (galactoside 2-alpha-L-fucosyltransferase), fucosyltransferase 1 (galactoside 2-alpha-L-fucosyltransferase, Bombayphenotype included), fucosyltransferase 1 (galactoside 2-alpha-L-fucosyltransferase, H blood group), galactoside 2-alpha-L-fucosyltransferase 1, GDP-L-fucose:beta-D-galactoside 2-alpha-L-fucosyltransferase 1, HHH, HSC | |
| Rabbit | |
| 41 kDa | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Canine: 100%; Goat: 100%; Equine: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Bovine: 92%; Guinea pig: 84%; Pig: 84%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Goat, Rabbit | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| P19526 | |
| FUT1 | |
| Synthetic peptides corresponding to FUT1 (fucosyltransferase 1 (galactoside 2-alpha-L-fucosyltransferase, H blood group)) The peptide sequence was selected from the middle region of FUT1 (NP_000139). Peptide sequence EATPWKDFALLTQCNHTIMTIGTFGFWAAYLAGGDTVYLANFTLPDSEFL The peptide sequence for this immunogen was taken from within the described region. | |
| Affinity purified | |
| RUO | |
| 2523 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction