missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Blood Group B Transferase/GTB/ABO Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | Blood Group B Transferase/GTB/ABO |
|---|---|
| Dilution | Western Blot 1.0 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
Blood Group B Transferase/GTB/ABO Polyclonal specifically detects Blood Group B Transferase/GTB/ABO in Mouse samples. It is validated for Western Blot.Specifications
| Blood Group B Transferase/GTB/ABO | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| A transferase, A3GALNT, A3GALT1, ABO blood group (transferase A, alpha 1-3-N-acetylgalactosaminyltransferase;transferase B, alpha 1-3-galactosyltransferase), ABO glycosyltransferase, B transferase, B(A) alpha-1,3-galactosyltransferase, EC 2.4.1.37, EC 2.4.1.40, Fucosylglycoprotein 3-alpha-galactosyltransferase, Fucosylglycoprotein alpha-N-acetylgalactosaminyltransferase, Glycoprotein-fucosylgalactoside alpha-galactosyltransferase, Glycoprotein-fucosylgalactoside alpha-N-acetylgalactosaminyltransferase, GTB, Histo-blood group A transferase, histo-blood group A2 transferase, histo-blood group ABO system transferase, Histo-blood group B transferase, NAGAT | |
| ABO | |
| IgG |
| Western Blot 1.0 ug/ml | |
| Polyclonal | |
| Rabbit | |
| P38649 | |
| 28 | |
| Synthetic peptides corresponding to the middle region of Abo. Immunizing peptide sequence GVEILSTFFGTLHPGFYSSSREAFTYERRPQSQAYIPWDRGDFYYGGAFF. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title