missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human BLK, His Tag Recombinant Protein
BLK plays an important role in B-cell activation.
Brand: Invitrogen™ PR6635A
Code : NEW
Additional Details : Weight : 0.01000kg
Description
B-lymphoid tyrosine kinase (BLK) is a non-receptor tyrosine kinase that belongs to the Src family of kinases.
Specifications
NP_001706 | |
62.3 kDa | |
Human | |
SRC Kinase Family | |
Recombinant | |
Storage Buffer: 50 mM Tris-HCl pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.05% Triton™ X-100, 4 mM DTT and 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Dry Ice | |
Kinases & Inhibitors | |
Liquid |
Kinase Assay | |
100μg | |
BLK, MGC10442 | |
Human | |
His Tag | |
Tyrosine Kinases | |
Baculovirus | |
Full Length, Recombinant, Cytoplasmic | |
Cell Cycle & Proliferation, Oncology |