missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human BLK, His Tag Recombinant Protein
BLK plays an important role in B-cell activation.
£434.00 - £6900.00
Specifications
Accession Number | NP_001706 |
---|---|
Gene Alias | BLK, MGC10442 |
KinaseFamily | SRC Kinase Family |
Species | Human |
Recombinant | Recombinant |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15448483
|
Invitrogen™
PR6635A |
100μg |
£1602.00
100µg |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10442523
|
Invitrogen™
PV4073 |
1mg |
£6900.00
1mg |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11563507
|
Invitrogen™
PV3683 |
10 μg |
£434.00
10µg |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
B-lymphoid tyrosine kinase (BLK) is a non-receptor tyrosine kinase that belongs to the Src family of kinases.
Specifications
NP_001706 | |
SRC Kinase Family | |
Recombinant | |
Baculovirus |
BLK, MGC10442 | |
Human | |
His Tag |