missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human BLK, His Tag Recombinant Protein
BLK plays an important role in B-cell activation.
Brand: Invitrogen™ PV3683
Code : 50
Additional Details : Weight : 0.05250kg
Description
B-lymphoid tyrosine kinase (BLK) is a non-receptor tyrosine kinase that belongs to the Src family of kinases. BLK plays an important role in B-cell activation.Specifications
NP_001706 | |
3554, 640 | |
10 μg | |
SRC Kinase Family | |
Recombinant | |
Storage Buffer: 50 mM Tris-HCl pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.05% Triton™ X-100, 4 mM DTT and 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Tyrosine Kinases | |
LanthaScreen™ Antibody, LanthaScreen™ Substrate, Z'-LYTE™, Omnia™ Bulk Substrate | |
B lymphoid tyrosine kinase | |
Kinases & Inhibitors | |
Liquid | |
Cell Cycle & Proliferation, Oncology |
Kinase Assay | |
62.3 kDa | |
BLK, MGC10442 | |
Human | |
His Tag | |
Biochemical phospho-specific activity (no antibody), Biochemical terbium phospho-specific activity, Biochemical binding/kinetic | |
Dry Ice | |
BLK | |
Baculovirus | |
Full Length, Recombinant, Cytoplasmic | |
Cytoplasmic Tyrosine Kinases |