missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human BLK, His Tag Recombinant Protein
BLK plays an important role in B-cell activation.
Brand: Invitrogen™ PV4073
Code : NEW
Additional Details : Weight : 0.00900kg
Description
B-lymphoid tyrosine kinase (BLK) is a non-receptor tyrosine kinase that belongs to the Src family of kinases.
Specifications
NP_001706 | |
50mM Tris-HCl pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.05% Triton™ X-100, 4mM DTT and 50% glycerol | |
1mg | |
RUO - research use only | |
BLK, MGC10442 | |
Human | |
His Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
62.3kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
SRC Kinase Family | |
Recombinant | |
Baculovirus |