missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Black Hanging Format Automation Pipet Tips for Hamilton Microlab™ Star Line and Nimbus™, 5 Pack
GREENER_CHOICEBIOPHARMA

Product Code. 15581403 Shop All Axygen Scientific Products
Click to view available options
Volume (Metric):
10 μL
50 μL
300 μL
1000 μL
Sterility:
Non-sterile
Sterile
Filtered:
Filtered
Non-filtered
Unit Size:
Pack of 3840
Pack of 5760
This item is not returnable. View return policy

Product Code. 15581403

Brand: Axygen™ HT50CBKHTRS5

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Compatible with Hamilton Microlab™ Star Line and Nimbus™ automated liquid handling workstations

Axygen offers an alternative choice for your Hamilton™ STARline system.

These tips are:

  • Manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality
  • Available in non-sterile, sterile, nonfiltered, and filtered
  • Available as Clear or Conductive black polypropylene tips
  • Available in 96 well hanging tip rack, nested tip rack, and individual deck-seating rack format
  • Designed for Hamilton™ MICROLAB™ STAR, STARlet, STARplus and Nimbus™ workstations
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 50 μL
Filtered Non-filtered
Sterility Sterile
Color Black
Autoclavable Not Autoclavable
Clean Claims DNase, RNase and Pyrogen free
For Use With (Application) Available in four sizes and specially designed for use with Hamilton™ liquid handling platforms, they enable precision pipetting across a wide range of applications
Format Hanging Tip Rack
Tip Style Automation
Packaging 5 Racks/Pk, 12 Pks/Cs.
For Use With (Equipment) Hamilton MICROLAB Nimbus
Material Polypropylene
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.