Learn More
Brady™ Black 7940 Series Thermal Transfer Printer Ribbon
Ribbon for Brady Printer BBP11 and BBP12
Brand:  Brady™ 804460
Additional Details : Weight : 0.07400kg
 View more versions of this product
Product Code. 11548814
  • £33.95 / 1 box
Estimated Shipment: 04-06-2024
to see stock.
Add to basket
Description
Description
  • General purpose, standard performance wax ribbon for paper and temporary labelling
Specifications
Specifications
Black
85 mm
Black 7940 Series Thermal Transfer Printer Ribbon
1 Ea.
70 m
12.7 mm I.D.
Ink side out