missing translation for 'onlineSavingsMsg'
Learn More

Bisoprolol Hemifumarate, TRC

Product Code. 17921875
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17921875 25 mg 25mg
17993835 50 mg 50mg
17924505 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17921875 Supplier Toronto Research Chemicals Supplier No. TRCB51050025MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104344-23-2
Molecular Formula 2 C18 H31 N O4 . C4 H4 O4
Molecular Weight (g/mol) 766.9582
Synonym 2-Propanol, 1-[4-[[2-(1-methylethoxy)ethoxy]methyl]phenoxy]-3-[(1-methylethyl)amino]-, (2E)-2-butenedioate (2:1) (salt) (9CI), 2-Propanol, 1-[4-[[2-(1-methylethoxy)ethoxy]methyl]phenoxy]-3-[(1-methylethyl)amino]-, (E)-2-butenedioate (2:1) (salt), 2-Propanol, 1-[4-[[2-(1-methylethoxy)ethoxy]methyl]phenoxy]-3-[(1-methylethyl)amino]-, (±)-, (E)-2-butenedioate (2:1) (salt) (8CI), Bisoprolol fumarate, Bisoprolol hemifumarate, Concor, Detensiel, EMD 33512, Emcor, Emvoncor, Eurtadal, Isoten, Maintate, Monocor, Soprol, Zebeta
IUPAC Name (E)-but-2-enedioic acid;1-(propan-2-ylamino)-3-[4-(2-propan-2-yloxyethoxymethyl)phenoxy]propan-2-ol
SMILES CC(C)NCC(O)COc1ccc(COCCOC(C)C)cc1.CC(C)NCC(O)COc2ccc(COCCOC(C)C)cc2.OC(=O)\C=C\C(=O)O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/2C18H31NO4.C4H4O4/c2*1-14(2)19-11-17(20)13-23-18-7-5-16(6-8-18)12-21-9-10-22-15(3)4;5-3(6)1-2-4(7)8/h2*5-8,14-15,17,19-20H,9-13H2,1-4H3;1-2H,(H,5,6)(H,7,8)/b;;2-1+
Formula Weight 766.4616
Percent Purity >95%
Chemical Name or Material Bisoprolol fumarate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.