missing translation for 'onlineSavingsMsg'
Learn More

Bisoprolol hemifumarate salt, 98%

Product Code. 15412438
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15412438 25 mg 25mg
15422438 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15412438 Supplier Thermo Scientific Alfa Aesar Supplier No. H56014.MB

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 104344-23-2 | C40H66N2O12 | 766.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104344-23-2
Molecular Formula C40H66N2O12
Molecular Weight (g/mol) 766.97
MDL Number MFCD00792743,MFCD00792743,MFCD00792743,MFCD00865795
InChI Key VMDFASMUILANOL-WXXKFALUNA-N
Synonym bisoprolol fumarate, 1-4-2-isopropoxyethoxy methyl phenoxy-3-isopropylamino propan-2-ol fumarate, but-2-enedioic acid; 1-propan-2-ylamino-3-4-2-propan-2-yloxyethoxymethyl phenoxy-2-propanol, but-2-enedioic acid; 1-propan-2-ylamino-3-4-2-propan-2-yloxyethoxymethyl phenoxy propan-2-ol
PubChem CID 53394791
IUPAC Name but-2-enedioic acid;1-(propan-2-ylamino)-3-[4-(2-propan-2-yloxyethoxymethyl)phenoxy]propan-2-ol
SMILES OC(=O)\C=C\C(O)=O.CC(C)NCC(O)COC1=CC=C(COCCOC(C)C)C=C1.CC(C)NCC(O)COC1=CC=C(COCCOC(C)C)C=C1

Specifications

Quantity 25 mg
Merck Index 14,1295
Formula Weight 383.49
Percent Purity 98%
Chemical Name or Material Bisoprolol hemifumarate salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.