missing translation for 'onlineSavingsMsg'
Learn More

Bisoprolol hemifumarate salt, 98%

Product Code. 15422438
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 15422438

Brand: Thermo Scientific Alfa Aesar H56014.MC

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 104344-23-2 | C40H66N2O12 | 766.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104344-23-2
Molecular Formula C40H66N2O12
Molecular Weight (g/mol) 766.97
MDL Number MFCD00792743,MFCD00792743,MFCD00792743,MFCD00865795
InChI Key VMDFASMUILANOL-WXXKFALUNA-N
Synonym bisoprolol fumarate, 1-4-2-isopropoxyethoxy methyl phenoxy-3-isopropylamino propan-2-ol fumarate, but-2-enedioic acid; 1-propan-2-ylamino-3-4-2-propan-2-yloxyethoxymethyl phenoxy-2-propanol, but-2-enedioic acid; 1-propan-2-ylamino-3-4-2-propan-2-yloxyethoxymethyl phenoxy propan-2-ol
PubChem CID 53394791
IUPAC Name but-2-enedioic acid;1-(propan-2-ylamino)-3-[4-(2-propan-2-yloxyethoxymethyl)phenoxy]propan-2-ol
SMILES OC(=O)\C=C\C(O)=O.CC(C)NCC(O)COC1=CC=C(COCCOC(C)C)C=C1.CC(C)NCC(O)COC1=CC=C(COCCOC(C)C)C=C1

Specifications

Quantity 100 mg
Merck Index 14,1295
Formula Weight 383.49
Percent Purity 98%
Chemical Name or Material Bisoprolol hemifumarate salt

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.