missing translation for 'onlineSavingsMsg'
Learn More

Bismarck Brown Y

Product Code. 11412668
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11412668 100 g 100g
11402668 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11412668 Supplier Thermo Scientific Alfa Aesar Supplier No. A16674.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10114-58-6 | C18H20Cl2N8 | 419.314 g/mol

Plasma stain for mucins, cartilage and goblet cellsBismarck Brown Y is used for staining mucine, cartilage and goblet cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Plasma stain for mucins, cartilage and goblet cellsBismarck Brown Y is used for staining mucine, cartilage and goblet cells.

Solubility
Soluble in water. Slightly soluble in ethanol and cellosolve. Insoluble in acetone, carbon tetrachloride and benzene.

Notes
Incompatible with strong oxidizing agents and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10114-58-6
Molecular Formula C18H20Cl2N8
Molecular Weight (g/mol) 419.314
MDL Number MFCD00012977
InChI Key MCZVRBLCRZWFJH-UHFFFAOYSA-N
Synonym C.I. 21000
PubChem CID 82360
ChEBI CHEBI:53615
IUPAC Name 4-[[3-[(2,4-diaminophenyl)diazenyl]phenyl]diazenyl]benzene-1,3-diamine;dihydrochloride
SMILES C1=CC(=CC(=C1)N=NC2=C(C=C(C=C2)N)N)N=NC3=C(C=C(C=C3)N)N.Cl.Cl

Specifications

Merck Index 14,1253
Solubility Information Soluble in water; Slightly soluble in ethanol and cellosolve; Insoluble in acetone,carbon tetrachloride and benzene.
Formula Weight 419.32
Odor Odorless
Quantity 100 g
Chemical Name or Material Bismarck Brown Y

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.