missing translation for 'onlineSavingsMsg'
Learn More

Bis(diphenylphosphino)acetylene, 98%, Thermo Scientific™

Product Code. 10787592 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10787592 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10787592 Supplier Thermo Scientific Acros Supplier No. 347790010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5112-95-8
Molecular Formula C26H20P2
Molecular Weight (g/mol) 394.39
MDL Number MFCD00003045
InChI Key FOWZHJNBFVLJGP-UHFFFAOYSA-N
Synonym bis diphenylphosphino acetylene, phosphine, ethynylenebis diphenyl, ethynylenebis diphenylphosphine, phosphine, 1,2-ethynediylbis diphenyl, 1,2-bis diphenylphosphino ethyne, 2-diphenylphosphanylethynyl diphenyl phosphane, 2-diphenylphosphanyl ethynyl diphenylphosphane, ethyne-1,2-diylbis diphenylphosphane, acmc-20almm, pubchem6917
PubChem CID 78789
IUPAC Name [2-(diphenylphosphanyl)ethynyl]diphenylphosphane
SMILES C1=CC=C(C=C1)P(C#CP(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 84.0°C to 88.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (C6H5)2PC≡CP(C6H5)2
Quantity 1 g
Formula Weight 394.39
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Bis(diphenylphosphino)acetylene, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Bis(diphenylphosphino)acetylene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.