accessibility menu, dialog, popup

UserName

CAS RN 5112-95-8

PP

CAS RN 5112-95-8

IUPAC Name: [2-(diphenylphosphanyl)ethynyl]diphenylphosphane
Synonyms: [2-(diphenylphosphanyl)ethynyl]diphenylphosphane
Molecular Weight (g/mol): 394.39
Molecular Formula: C26H20P2
InChi Key: FOWZHJNBFVLJGP-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)P(C#CP(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)