missing translation for 'onlineSavingsMsg'
Learn More

Bis(dimethylamino)chlorophosphine

Product Code. 11397248
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11397248 1 g 1g
11387248 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11397248 Supplier Thermo Scientific Alfa Aesar Supplier No. 030307.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 3348-44-5 | C4H12ClN2P | 154.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3348-44-5
Molecular Formula C4H12ClN2P
Molecular Weight (g/mol) 154.58
MDL Number MFCD00014862
InChI Key MAJFLEHNBOUSIY-UHFFFAOYSA-N
Synonym bis dimethylamino chlorophosphine, chlorobis dimethylamino phosphine, phosphorodiamidous chloride, tetramethyl, chloro dimethylamino phosphanyl dimethylamine, p nme2 2cl, acmc-1cr19, ch3 2n 2pcl, tetramethyldiaminochlorophosphine, bis dimethylamino phosphoruschloride, bis dimethylamino phosphorus chloride
PubChem CID 76872
IUPAC Name N-[chloro(dimethylamino)phosphanyl]-N-methylmethanamine
SMILES CN(C)P(Cl)N(C)C

Specifications

Color Colorless
Odor Pungent
Linear Formula [(CH3)2N]2PCl
Refractive Index 1.501
Quantity 1 g
UN Number UN3265
Sensitivity Air and moisture sensitive
Formula Weight 154.58
Physical Form Liquid
Chemical Name or Material Bis(dimethylamino)chlorophosphine
compliance-icons
Product Identifier
  • Bis(dimethylamino)chlorophosphine
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308-IF exposed or concerned:.
  • P402-Store in a dry place.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.