accessibility menu, dialog, popup

UserName

CH3NH3CPClNCH3H3C

CAS RN 3348-44-5

IUPAC Name: [chloro(dimethylamino)phosphanyl]dimethylamine
Synonyms: [chloro(dimethylamino)phosphanyl]dimethylamine
Molecular Weight (g/mol): 154.58
Molecular Formula: C4H12ClN2P
InChi Key: MAJFLEHNBOUSIY-UHFFFAOYSA-N
SMILES: CN(C)P(Cl)N(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results