missing translation for 'onlineSavingsMsg'
Learn More

Bis[2-(diisopropylphosphino)ethyl]amine, 10% w/w soln. in THF

Product Code. 15455776
Change view
Click to view available options
Quantity:
1 g
25 g
Unit Size:
1g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15455776 1 g 1g
15475776 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15455776 Supplier Thermo Scientific Alfa Aesar Supplier No. H60468.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 131890-26-1 | C16H37NP2 | 305.427 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis[2-(diisopropylphosphino)ethyl]amine acts as ligand and form complexes with ruthenium, which finds application in the dehydrogenation of ethanol.

Solubility
Immiscible with water.

Notes
Air sensitive. Incompatible with oxidizing agents and oxygen.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 131890-26-1
Molecular Formula C16H37NP2
Molecular Weight (g/mol) 305.427
MDL Number MFCD17018778
InChI Key FTVIGQGOGIHMBS-UHFFFAOYSA-N
Synonym bis 2-di-i-propylphosphino ethyl amine, di 2-diisopropylphosphino ethyl amine, bis 2-diisopropylphosphino ethyl amine, bis 2-diisopropylphosphanyl ethyl amine, bis 2-diisopropylphosphino ethyl amine solution, 10 wt. % in thf
PubChem CID 15757160
IUPAC Name 2-di(propan-2-yl)phosphanyl-N-[2-di(propan-2-yl)phosphanylethyl]ethanamine
SMILES CC(C)P(CCNCCP(C(C)C)C(C)C)C(C)C

Specifications

Density 0.884
Linear Formula ([(CH3)2CH]2PCH2CH2)2NH
Refractive Index 1.418
Quantity 1 g
UN Number UN2924
Sensitivity Air Sensitive
Solubility Information Immiscible with water.
Formula Weight 305.42
Concentration or Composition (by Analyte or Components) 10% w/w soln. in THF
Chemical Name or Material Bis[2-(diisopropylphosphino)ethyl]amine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.