accessibility menu, dialog, popup

UserName

CAS RN 131890-26-1

CH3CH3PHNPH3CCH3CH3H3CCH3H3C

CAS RN 131890-26-1

IUPAC Name: bis({2-[bis(propan-2-yl)phosphanyl]ethyl})amine
Synonyms: bis[2-(diisopropylphosphanyl)ethyl]amine
Molecular Weight (g/mol): 305.43
Molecular Formula: C16H37NP2
InChi Key: FTVIGQGOGIHMBS-UHFFFAOYSA-N
SMILES: CC(C)P(CCNCCP(C(C)C)C(C)C)C(C)C
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results