missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Biotinyl-Ala-Ser-Thr-DL-Asp fluoromethyl ketone

Product Code. 15456289
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15456289 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15456289 Supplier Thermo Scientific™ Supplier No. J64265.M^

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A general caspase inhibitor

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 mg
Description White to off-white
Content And Storage -30°C to -10°C
For Use With (Application) A general caspase inhibitor
Molecular Formula C25H39FN6O10S
Formula Weight 634.67
Synonym Biotinyl-ASTD-FMK;
Product Type Biotinyl-Ala-Ser-Thr-DL-Asp fluoromethyl ketone
Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.